Assay Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112716.1 | NP_001106187.1 | 8 - 9 | 1344 | 89 | |
| NM_181373.3 | NP_852038.2 | 8 - 9 | 1344 | 89 | ||
| XM_006238857.2 | XP_006238919.1 | 8 - 9 | 1935 | 89 | ||
| GenBank mRNA | AF027331.1 | - | 8 - 9 | 1347 | 89 | |
| M83552.1 | - | 8 - 9 | 1120 | 89 | ||
| Z11716.1 | - | 8 - 9 | 1347 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
glutamate receptor signaling pathway
G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of membrane potential
negative regulation of synaptic transmission, glutamatergic
adenylate cyclase inhibiting G-protein coupled glutamate receptor activity
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
kainate selective glutamate receptor activity
signal transduction
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
glutamate receptor signaling pathway
G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of membrane potential
negative regulation of synaptic transmission, glutamatergic
adenylate cyclase inhibiting G-protein coupled glutamate receptor activity
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
kainate selective glutamate receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36075 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042514 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042514 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042514 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P242904 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58710 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85332 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77815 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16696 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11868 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99112 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29821 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72142 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77971 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30866 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24739 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93173 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13805 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47450 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37849 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3581 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3895 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69641 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10871943 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871945 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871956 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871942 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871954 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871955 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10871939 | - | - |
| Rat Genome 230 2.0 Array | 1370587_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370587_at | - | - |
| Affymetrix HT Rat Focus Array | 1370587_at | - | - |
| Affymetrix HT Rat Focus Array | 1370587_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500010193.rn.1 | TC0500001450.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370587_PM_at | - | - |
| Rat Expression Set 230 | 1370587_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871939 | - | - |
| Rat Genome U34 Set | AF027331_at | - | - |
| Rat Genome U34 Set | AF027331_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21734158 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352169 | - | - |

Grik3
Genomic Map