Assay Details
| Gene Symbol: Osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106716.2 | NP_001100186.1 | 1 - 2 | 710 | 75 | |
| XM_006239886.2 | XP_006239948.1 | 2 - 3 | 1004 | 75 | ||
| GenBank mRNA | BC159410.1 | - | 1 - 2 | 710 | 8366695 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Osr1
Genomic Map