Assay Details
| Gene Symbol: Aldh1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017272.15 | NP_058968.14 | 10 - 11 | 1403 | 80 | |
| XM_008760315.1 | XP_008758537.1 | 10 - 11 | 1382 | 80 | ||
| GenBank mRNA | M23995.1 | - | 10 - 11 | 1403 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ethanol catabolic process
operant conditioning
protein homotetramerization
oxidation-reduction process
retinal dehydrogenase activity
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
benzaldehyde dehydrogenase (NAD+) activity
identical protein binding
ethanol catabolic process
operant conditioning
protein homotetramerization
oxidation-reduction process
retinal dehydrogenase activity
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
benzaldehyde dehydrogenase (NAD+) activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P335974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72409 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P335974 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P335974 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P335974 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P335974 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701521645 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701234495 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78203 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1168 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14245 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 387 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701234495 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701521645 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10729287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714330 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729286 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10714323 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10729284 | - | - |
| Rat Genome 230 2.0 Array | 1368718_at | - | - |
| Rat Genome 230 2.0 Array | 1368718_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368718_at | - | - |
| Affymetrix HT Rat Focus Array | 1368718_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100059877.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059880.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059883.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059864.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059862.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059863.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059871.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059878.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059865.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059866.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059882.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059884.rn.1 | TC0100007414.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059881.rn.1 | TC0100007414.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368718_PM_at | - | - |
| Rat Expression Set 230 | 1368718_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714323 | - | - |
| Rat Genome U34 Set | M23995_g_at | - | - |
| Rat Genome U34 Set | M23995_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21273026 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371599 | - | - |
| RatRef-12 Expression BeadChip | ILMN_55820 | - | - |

Aldh1a7
Genomic Map