Assay Details
| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270983.1 | NP_001257912.1 | 6 - 7 | 1136 | 81 | |
| NM_022498.2 | NP_071943.1 | 6 - 7 | 1136 | 81 | ||
| XM_008769212.1 | XP_008767434.1 | 7 - 8 | 1023 | 81 | ||
| GenBank mRNA | BC078825.1 | - | 6 - 7 | 1063 | 7112668 | 81 |
| D90165.1 | - | 6 - 7 | 965 | 81 | ||
| D90166.1 | - | 6 - 7 | 1079 | 81 | ||
| FM122849.1 | - | 3 - 4 | 448 | 81 | ||
| FQ228186.1 | - | 2 - 3 | 250 | 81 | ||
| FQ232892.1 | - | 2 - 3 | 250 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1cc
Genomic Map