Assay Details
| Gene Symbol: Ces1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031565.2 | NP_113753.2 | 8 - 9 | 1278 | 87 | |
| GenBank mRNA | FM062488.1 | - | 5 - 6 | 402 | 87 | |
| X81395.1 | - | 8 - 9 | 1143 | 87 | ||
| Gene Symbol: LOC102557406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772338.1 | XP_008770560.1 | 1 - 2 | 570 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ces1e
Genomic Map