Assay Details
| Gene Symbol: Nr2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080778.2 | NP_542956.1 | 1 - 2 | 628 | 79 | |
| GenBank mRNA | AF003944.1 | - | 1 - 2 | 629 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr2f2
Genomic Map