Assay Details
| Gene Symbol: Tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145778.2 | NP_665721.1 | 3 - 4 | 447 | 69 | |
| GenBank mRNA | AB015946.1 | - | 3 - 4 | 352 | 69 | |
| BC070957.1 | - | 3 - 4 | 447 | 7105643 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubg1
Genomic Map