Assay Details
| Gene Symbol: Hnrnpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175603.2 | NP_783193.2 | 2 - 3 | 238 | 87 | |
| GenBank mRNA | AY184814.1 | - | 1 - 2 | 152 | 87 | |
| BC093598.1 | - | 2 - 3 | 238 | 7315896 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P048133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31154 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P048133 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P048133 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031025 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P048133 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P840829 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P210235 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2378 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24465 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44725 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10872997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10872999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10872995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10872996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10872998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873004 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10872994 | - | - |
| Rat Genome 230 2.0 Array | 1373905_at | - | - |
| Rat Genome 230 2.0 Array | 1373905_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373905_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373905_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500012438.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012443.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012433.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012445.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012441.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012430.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012447.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012434.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012439.rn.1 | TC0500001704.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012440.rn.1 | TC0500001704.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373905_at | - | - |
| Rat Expression Set 230 | 1373905_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10872994 | - | - |
| Rat Genome U34 Set | rc_AI178441_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22271255 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366710 | - | - |
| RatRef-12 Expression BeadChip | ILMN_47875 | - | - |

Hnrnpr
Genomic Map