Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270844.1 | NP_001257773.1 | 7 - 8 | 1003 | 135 | |
| NM_001270845.1 | NP_001257774.1 | 7 - 8 | 925 | 135 | ||
| NM_182815.2 | NP_877967.2 | 8 - 9 | 1120 | 135 | ||
| XM_006249713.2 | XP_006249775.1 | 8 - 9 | 1213 | 135 | ||
| XM_008769436.1 | XP_008767658.1 | 7 - 8 | 1135 | 135 | ||
| XM_008769437.1 | XP_008767659.1 | 8 - 9 | 1136 | 135 | ||
| XM_008769438.1 | XP_008767660.1 | 8 - 9 | 1119 | 135 | ||
| XM_008769439.1 | XP_008767661.1 | 7 - 8 | 1019 | 135 | ||
| XM_008769440.1 | XP_008767662.1 | 8 - 9 | 1306 | 135 | ||
| GenBank mRNA | AY278363.1 | - | 8 - 9 | 1120 | 135 | |
| AY278364.1 | - | 7 - 8 | 925 | 135 | ||
| BC079287.1 | - | 7 - 8 | 982 | 7134311 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zp3r
Genomic Map