Assay Details
| Gene Symbol: Myrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182844.1 | NP_878264.1 | 8 - 9 | 1022 | 101 | |
| XM_006244110.2 | XP_006244172.1 | 8 - 9 | 1062 | 101 | ||
| XM_008766684.1 | XP_008764906.1 | 9 - 10 | 1140 | 101 | ||
| XM_008766685.1 | XP_008764907.1 | 10 - 11 | 1254 | 101 | ||
| XM_008766686.1 | XP_008764908.1 | 8 - 9 | 1064 | 101 | ||
| XM_008766687.1 | XP_008764909.1 | 7 - 8 | 1535 | 101 | ||
| GenBank mRNA | AY331984.1 | - | 8 - 9 | 1022 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P392359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94054 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P392359 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P392359 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P392359 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P392359 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99204 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92576 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13161 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6627 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63106 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1241 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70243 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58142 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10930 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93370 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90262 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58306 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10914338 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914340 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914333 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914337 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914334 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914336 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914339 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914332 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914341 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914335 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914345 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10914331 | - | - |
| Rat Genome 230 2.0 Array | 1384095_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373328_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384095_at | - | - |
| Rat Genome 230 2.0 Array | 1380826_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384183_at | - | - |
| Rat Genome 230 2.0 Array | 1373328_at | - | - |
| Rat Genome 230 2.0 Array | 1380826_at | - | - |
| Rat Genome 230 2.0 Array | 1384183_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384095_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384095_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800014043.rn.1 | TC0800001761.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014044.rn.1 | TC0800001761.rn.1 | RTA TC |
| Rat Expression Set 230 | 1380826_PM_at | - | - |
| Rat Expression Set 230 | 1384095_PM_at | - | - |
| Rat Expression Set 230 | 1384095_at | - | - |
| Rat Expression Set 230 | 1380826_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914331 | - | - |
| Rat Genome U34 Set | rc_AI229482_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21323218 | - | - |
| ABI Rat Genome Survey Microarray | 21615622 | - | - |
| ABI Rat Genome Survey Microarray | 21746457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370776 | - | - |
| RatRef-12 Expression BeadChip | ILMN_50156 | - | - |

Myrip
Genomic Map