Assay Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107773.1 | NP_001101243.1 | 2 - 3 | 1953 | 85 | |
| XM_006234974.2 | XP_006235036.1 | 2 - 3 | 901 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk35
Genomic Map