Assay Details
| Gene Symbol: Ncl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012749.2 | NP_036881.2 | 9 - 10 | 1587 | 86 | |
| GenBank mRNA | BC085751.1 | - | 9 - 10 | 1587 | 7109097 | 86 |
| FJ817497.1 | - | 9 - 10 | 1026 | 86 | ||
| FQ219308.1 | - | 2 - 3 | 199 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ncl
Genomic Map