Assay Details
| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013053.1 | NP_037185.1 | 3 - 4 | 497 | 71 | |
| GenBank mRNA | BC062409.1 | - | 2 - 3 | 467 | 5623850 | 71 |
| D17614.1 | - | 3 - 4 | 497 | 71 | ||
| FQ218679.1 | - | 2 - 3 | 257 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ywhaq
Genomic Map