Assay Details
| Gene Symbol: Hsd3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007719.3 | NP_001007720.3 | 2 - 3 | 295 | 62 | |
| GenBank mRNA | BC086578.1 | - | 2 - 3 | 295 | 7306702 | 62 |
| M38178.1 | - | 2 - 3 | 314 | 62 | ||
| Gene Symbol: Hsd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042619.1 | NP_001036084.1 | 2 - 3 | 295 | 62 | |
| GenBank mRNA | M38179.1 | - | 2 - 3 | 295 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Hsd3b1
Genomic Map