Assay Details
| Gene Symbol: Timm23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019352.2 | NP_062225.1 | 2 - 3 | 268 | 107 | |
| GenBank mRNA | AB006451.1 | - | 2 - 3 | 268 | 107 | |
| BC083559.1 | - | 2 - 3 | 275 | 7135081 | 107 | |
| FQ216304.1 | - | 2 - 3 | 272 | 107 | ||
| FQ229692.1 | - | 2 - 3 | 272 | 107 | ||
| FQ229803.1 | - | 2 - 3 | 274 | 107 | ||
| Gene Symbol: LOC100362432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729199.4 | XP_002729245.1 | 1 | 381 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Timm23
Genomic Map