Assay Details
| Gene Symbol: Rpl36al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031105.2 | NP_112367.1 | 2 | 371 | 65 | |
| GenBank mRNA | AA955285.1 | - | 65 | |||
| BC058142.1 | - | 374 | 6919511 | 65 | ||
| FQ218934.1 | - | 2 | 371 | 65 | ||
| M19635.1 | - | 325 | 65 | |||
| Gene Symbol: LOC100912182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751738.3 | XP_003751786.1 | 2 | 397 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl36al
Genomic Map