Assay Details
| Gene Symbol: Rps12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031709.3 | NP_113897.2 | 3 - 4 | 249 | 56 | |
| GenBank mRNA | BC058460.1 | - | 3 - 4 | 253 | 6919500 | 56 |
| FQ210032.1 | - | 3 - 4 | 214 | 56 | ||
| FQ212402.1 | - | 3 - 4 | 212 | 56 | ||
| FQ212636.1 | - | 3 - 4 | 213 | 56 | ||
| FQ217474.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221068.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221108.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221199.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221263.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221353.1 | - | 3 - 4 | 215 | 56 | ||
| FQ221368.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221420.1 | - | 3 - 4 | 213 | 56 | ||
| FQ221550.1 | - | 3 - 4 | 214 | 56 | ||
| FQ221711.1 | - | 3 - 4 | 215 | 56 | ||
| FQ222260.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222326.1 | - | 3 - 4 | 211 | 56 | ||
| FQ222422.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222486.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222505.1 | - | 3 - 4 | 214 | 56 | ||
| FQ222591.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222620.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222621.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222742.1 | - | 3 - 4 | 216 | 56 | ||
| FQ222863.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222888.1 | - | 3 - 4 | 213 | 56 | ||
| FQ222895.1 | - | 3 - 4 | 213 | 56 | ||
| FQ223029.1 | - | 3 - 4 | 214 | 56 | ||
| FQ223134.1 | - | 3 - 4 | 214 | 56 | ||
| FQ223216.1 | - | 3 - 4 | 213 | 56 | ||
| FQ223305.1 | - | 3 - 4 | 216 | 56 | ||
| FQ223374.1 | - | 3 - 4 | 216 | 56 | ||
| FQ223466.1 | - | 3 - 4 | 214 | 56 | ||
| FQ228598.1 | - | 3 - 4 | 213 | 56 | ||
| FQ228992.1 | - | 3 - 4 | 214 | 56 | ||
| FQ230145.1 | - | 3 - 4 | 213 | 56 | ||
| M18547.1 | - | 3 - 4 | 212 | 56 | ||
| Gene Symbol: LOC100360573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728597.4 | XP_002728643.1 | 1 - 2 | 213 | 56 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps12
Genomic Map