Assay Details
| Gene Symbol: Sumo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133594.2 | NP_598278.1 | 1 - 2 | 181 | 101 | |
| XM_008768397.1 | XP_008766619.1 | 2 - 3 | 321 | 107 | ||
| GenBank mRNA | BC058446.1 | - | 144 | 6921523 | 101 | |
| BC078746.1 | - | 1 - 2 | 115 | 7125132 | 101 | |
| CF976332.1 | - | 185 | 101 | |||
| FQ221996.1 | - | 148 | 101 | |||
| FQ225379.1 | - | 123 | 101 | |||
| FQ225921.1 | - | 139 | 101 | |||
| FQ225989.1 | - | 139 | 101 | |||
| FQ230134.1 | - | 150 | 101 | |||
| FQ235035.1 | - | 1 - 2 | 62 | 101 | ||
| L79949.1 | - | 1 - 2 | 128 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058446.1,BC078746.1,L79949.1,NM_133594.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sumo2
Genomic Map