Assay Details
| Gene Symbol: Gabarap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172036.3 | NP_742033.1 | 2 - 3 | 220 | 101 | |
| GenBank mRNA | AF161588.1 | - | 2 - 3 | 234 | 101 | |
| BC058441.1 | - | 2 - 3 | 224 | 6918795 | 101 | |
| FQ213199.1 | - | 2 - 3 | 291 | 101 | ||
| FQ224284.1 | - | 2 - 3 | 186 | 101 | ||
| FQ229256.1 | - | 2 - 3 | 222 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gabarap
Genomic Map