Assay Details
| Gene Symbol: Tpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173111.1 | NP_775134.1 | 4 - 5 | 522 | 72 | |
| XM_006232568.2 | XP_006232630.1 | 5 - 6 | 676 | 72 | ||
| XM_006232569.2 | XP_006232631.1 | 5 - 6 | 676 | 72 | ||
| XM_006232570.2 | XP_006232632.1 | 5 - 6 | 676 | 72 | ||
| XM_006232571.2 | XP_006232633.1 | 5 - 6 | 676 | 72 | ||
| XM_006232576.2 | XP_006232638.1 | 5 - 6 | 676 | 72 | ||
| XM_006232580.2 | XP_006232642.1 | 4 - 5 | 580 | 72 | ||
| XM_006232584.2 | XP_006232646.1 | 4 - 5 | 582 | 72 | ||
| XM_008761124.1 | XP_008759346.1 | 4 - 5 | 580 | 72 | ||
| XM_008761125.1 | XP_008759347.1 | 4 - 5 | 681 | 72 | ||
| XM_008761126.1 | XP_008759348.1 | 3 - 4 | 447 | 72 | ||
| GenBank mRNA | FQ228755.1 | - | 4 - 5 | 530 | 72 | |
| L24775.1 | - | 4 - 5 | 480 | 72 | ||
| S82383.1 | - | 4 - 5 | 499 | 72 | ||
| X72859.1 | - | 4 - 5 | 522 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P003937 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P129137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121091 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P003937 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P129137 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P129137 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P003937 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P003937 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P129137 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P219839 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P513058 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P134685 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P277834 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P121177 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P346661 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P134688 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18140 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1360 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66514 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10816890 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10934691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816899 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816897 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816889 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816898 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797727 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10816886 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10934689 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10797726 | - | - |
| Rat Genome 230 2.0 Array | 1387617_at | - | - |
| Rat Genome 230 2.0 Array | 1370450_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371239_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1371184_x_at | - | - |
| Rat Genome 230 2.0 Array | 1398303_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1388268_at | - | - |
| Rat Genome 230 2.0 Array | 1370450_at | - | - |
| Rat Genome 230 2.0 Array | 1387918_at | - | - |
| Rat Genome 230 2.0 Array | 1376929_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383986_at | - | - |
| Rat Genome 230 2.0 Array | 1370340_x_at | - | - |
| Rat Genome 230 2.0 Array | 1383986_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371239_s_at | - | - |
| Rat Genome 230 2.0 Array | 1370340_PM_x_at | - | - |
| Rat Genome 230 2.0 Array | 1387918_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388268_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389846_at | - | - |
| Rat Genome 230 2.0 Array | 1389846_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398303_s_at | - | - |
| Rat Genome 230 2.0 Array | 1387617_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371184_PM_x_at | - | - |
| Rat Genome 230 2.0 Array | 1376929_at | - | - |
| Affymetrix HT Rat Focus Array | 1398303_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1371184_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1387617_at | - | - |
| Affymetrix HT Rat Focus Array | 1376929_at | - | - |
| Affymetrix HT Rat Focus Array | 1371239_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1387918_at | - | - |
| Affymetrix HT Rat Focus Array | 1376929_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389846_at | - | - |
| Affymetrix HT Rat Focus Array | 1371239_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1370450_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370340_PM_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1371184_PM_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1387918_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370450_at | - | - |
| Affymetrix HT Rat Focus Array | 1389846_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398303_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1370340_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1387617_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200010532.rn.1 | TC0200001596.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200010538.rn.1 | TC0200001596.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200010522.rn.1 | TC0200001596.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370339_at | - | - |
| Rat Expression Set 230 | 1370450_PM_at | - | - |
| Rat Expression Set 230 | 1376929_at | - | - |
| Rat Expression Set 230 | 1387918_at | - | - |
| Rat Expression Set 230 | 1371239_PM_s_at | - | - |
| Rat Expression Set 230 | 1370339_PM_at | - | - |
| Rat Expression Set 230 | 1389846_at | - | - |
| Rat Expression Set 230 | 1398303_PM_s_at | - | - |
| Rat Expression Set 230 | 1376929_PM_at | - | - |
| Rat Expression Set 230 | 1388268_at | - | - |
| Rat Expression Set 230 | 1370450_at | - | - |
| Rat Expression Set 230 | 1371239_s_at | - | - |
| Rat Expression Set 230 | 1387617_at | - | - |
| Rat Expression Set 230 | 1388268_PM_at | - | - |
| Rat Expression Set 230 | 1370340_x_at | - | - |
| Rat Expression Set 230 | 1370340_PM_x_at | - | - |
| Rat Expression Set 230 | 1387617_PM_at | - | - |
| Rat Expression Set 230 | 1371184_PM_x_at | - | - |
| Rat Expression Set 230 | 1387918_PM_at | - | - |
| Rat Expression Set 230 | 1389846_PM_at | - | - |
| Rat Expression Set 230 | 1398303_s_at | - | - |
| Rat Expression Set 230 | 1371184_x_at | - | - |
| Rat Expression Set 230 | 1383986_at | - | - |
| Rat Expression Set 230 | 1383986_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10816886 | - | - |
| Rat Genome U34 Set | rc_AA859305_s_at | - | - |
| Rat Genome U34 Set | rc_AA818554_s_at | - | - |
| Rat Genome U34 Set | L24776_at | - | - |
| Rat Genome U34 Set | S82383_s_at | - | - |
| Rat Genome U34 Set | L24777_at | - | - |
| Rat Genome U34 Set | rc_AI071098_at | - | - |
| Rat Genome U34 Set | rc_AI170775_at | - | - |
| Rat Genome U34 Set | rc_AI170847_g_at | - | - |
| Rat Genome U34 Set | rc_AI170847_at | - | - |
| Rat Genome U34 Set | rc_AI179415_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20923949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363774 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649963 | - | - |
| RatRef-12 Expression BeadChip | ILMN_54877 | - | - |

Tpm3
Genomic Map