Assay Details
| Gene Symbol: Sult1c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013177.2 | NP_001013195.2 | 931 | 151 | ||
| GenBank mRNA | AJ238392.1 | - | 872 | 151 | ||
| BC081691.1 | - | 828 | 7110136 | 151 | ||
| BC092564.1 | - | 931 | 7372924 | 151 | ||
| FQ209913.1 | - | 925 | 151 | |||
| Gene Symbol: RGD1559960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244256.2 | XP_006244318.2 | 4 - 5 | 615 | 151 | |
| XM_008766792.1 | XP_008765014.1 | 5 - 6 | 762 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P141963 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P958682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128873 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108035 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P958682 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P141963 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P141963 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108035 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P958682 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P141963 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P958682 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P108035 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P211168 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P141963 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P958682 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701716372 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700539477 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700599447 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20951 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17476 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18029 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701716372 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700539477 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700599447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10921244 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926102 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921240 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921241 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926099 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921243 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921242 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921245 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926103 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10921227 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10921239 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10926125 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10926098 | - | - |
| Rat Genome 230 2.0 Array | 1370943_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370943_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900011640.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011635.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011638.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011636.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011639.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011653.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011658.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011660.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011637.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011654.rn.1 | TC0900001406.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900011655.rn.1 | TC0900001406.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370943_at | - | - |
| Rat Expression Set 230 | 1370943_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921227 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921239 | - | - |
| Rat Genome U34 Set | rc_AA926193_at | - | - |
| Rat Genome U34 Set | rc_AI638982_at | - | - |
| Rat Genome U34 Set | rc_AA926193_at | - | - |
| Rat Genome U34 Set | rc_AA926193_at | - | - |
| Rat Genome U34 Set | rc_AI168953_at | - | - |
| Rat Genome U34 Set | rc_AA926193_at | - | - |
| Rat Genome U34 Set | rc_AA926193_at | - | - |
| Rat Genome U34 Set | rc_AA926193_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20884839 | - | - |
| ABI Rat Genome Survey Microarray | 21106968 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367559 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375047 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66044 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66424 | - | - |

Sult1c2a
Genomic Map