Assay Details
| Gene Symbol: Inpp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108923.2 | NP_001102393.1 | 1 - 2 | 362 | 63 | |
| XM_006230486.2 | XP_006230548.1 | 1 - 2 | 669 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P040739 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P375137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162923 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P375137 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040739 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P375137 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040739 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P040739 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P375137 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P201981 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P463996 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60717 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58297 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73986 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81658 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10711962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711969 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711967 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711966 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711965 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711963 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711971 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10711961 | - | - |
| Rat Genome 230 2.0 Array | 1388697_at | - | - |
| Rat Genome 230 2.0 Array | 1388697_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390774_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390774_at | - | - |
| Affymetrix HT Rat Focus Array | 1388697_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388697_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100022252.rn.1 | TC0100002824.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100022257.rn.1 | TC0100002824.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100022253.rn.1 | TC0100002824.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100022250.rn.1 | TC0100002824.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388697_PM_at | - | - |
| Rat Expression Set 230 | 1388697_at | - | - |
| Rat Expression Set 230 | 1390774_at | - | - |
| Rat Expression Set 230 | 1390774_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711961 | - | - |
| Rat Genome U34 Set | rc_AA850544_at | - | - |
| Rat Genome U34 Set | rc_AA963131_i_at | - | - |
| Rat Genome U34 Set | rc_AA963131_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20754790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374350 | - | - |

Inpp5a
Genomic Map