Assay Details
| Gene Symbol: Camta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195559.1 | NP_001182488.1 | 169 | 95 | ||
| NM_001195560.1 | NP_001182489.1 | 2 - 3 | 169 | 95 | ||
| GenBank mRNA | BC106025.1 | - | 2 - 3 | 169 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P210667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39747 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P589657 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P210667 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P589657 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P210667 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P210667 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P589657 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80631 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101049 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3900 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10881919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881918 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10881917 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10881892 | - | - |
| Rat Genome 230 2.0 Array | 1389636_at | - | - |
| Rat Genome 230 2.0 Array | 1389636_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389636_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389636_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500031522.rn.1 | TC0500004118.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500031524.rn.1 | TC0500004118.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500031519.rn.1 | TC0500004118.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500031520.rn.1 | TC0500004118.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500031523.rn.1 | TC0500004118.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500031521.rn.1 | TC0500004118.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500031512.rn.1 | TC0500004118.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389636_at | - | - |
| Rat Expression Set 230 | 1389636_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881917 | - | - |
| Rat Genome U34 Set | rc_AI172230_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361124 | - | - |

Camta1
Genomic Map