Assay Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134760.1 | NP_001128232.1 | 2 - 3 | 370 | 69 | |
| NM_032619.2 | NP_116008.2 | 2 - 3 | 370 | 69 | ||
| XM_006228691.2 | XP_006228753.1 | 2 - 3 | 514 | 69 | ||
| XM_006228692.2 | XP_006228754.1 | 2 - 3 | 514 | 69 | ||
| GenBank mRNA | BC169048.1 | - | 2 - 3 | 325 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpl
Genomic Map