Assay Details
| Gene Symbol: Dpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105729.3 | NP_001099199.2 | 3 - 4 | 372 | 56 | |
| XM_006228677.2 | XP_006228739.1 | 3 - 4 | 409 | 56 | ||
| XM_006228678.2 | XP_006228740.1 | 3 - 4 | 406 | 56 | ||
| XM_006228679.2 | XP_006228741.1 | 3 - 4 | 406 | 56 | ||
| XM_006228680.2 | XP_006228742.1 | 3 - 4 | 411 | 56 | ||
| XM_006228681.2 | XP_006228743.1 | 3 - 4 | 407 | 56 | ||
| XM_006228682.2 | XP_006228744.1 | 3 - 4 | 413 | 56 | ||
| XM_006228684.2 | XP_006228746.1 | 3 - 4 | 1108 | 56 | ||
| XM_006228685.1 | XP_006228747.1 | 3 - 4 | 1233 | 56 | ||
| XM_008759180.1 | XP_008757402.1 | 3 - 4 | 395 | 56 | ||
| GenBank mRNA | BC159415.1 | - | 3 - 4 | 300 | 8368092 | 56 |
| CO396551.1 | - | 3 - 4 | 372 | 56 | ||
| X66022.1 | - | 3 - 4 | 1233 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpf1
Genomic Map