Assay Details
| Gene Symbol: Spa17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053482.1 | NP_445934.1 | 1 - 2 | 160 | 87 | |
| XM_006242812.2 | XP_006242874.1 | 2 - 3 | 336 | 87 | ||
| GenBank mRNA | AJ131888.1 | - | 1 - 2 | 160 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spa17
Genomic Map