Assay Details
| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013181.1 | NP_001013199.1 | 4 - 5 | 1612 | 64 | |
| GenBank mRNA | AB568265.1 | - | 3 - 4 | 410 | 64 | |
| AY781102.1 | - | 4 - 5 | 1612 | 64 | ||
| AY781103.1 | - | 4 - 5 | 1612 | 64 | ||
| AY781104.1 | - | 4 - 5 | 1603 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb16
Genomic Map