Assay Details
| Gene Symbol: Ythdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047099.1 | NP_001040564.1 | 2 - 3 | 184 | 133 | |
| GenBank mRNA | AB259152.1 | - | 2 - 3 | 184 | 133 | |
| BC166407.1 | - | 1 - 2 | 99 | 7453874 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB259152.1, BC166407.1, NM_001047099.1 transcripts.
Set Membership: |
|

Ythdf2
Genomic Map