Assay Details
| Gene Symbol: Ythdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047099.1 | NP_001040564.1 | 3 - 4 | 211 | 112 | |
| GenBank mRNA | AB259152.1 | - | 3 - 4 | 211 | 112 | |
| BC166407.1 | - | 2 - 3 | 126 | 7453874 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ythdf2
Genomic Map