Assay Details
| Gene Symbol: Epm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276762.1 | NP_001263691.1 | 1 - 2 | 305 | 88 | |
| GenBank mRNA | AF347030.1 | - | 293 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epm2a
Genomic Map