Assay Details
| Gene Symbol: Mycbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271324.1 | NP_001258253.1 | 1 - 2 | 88 | 67 | |
| GenBank mRNA | FM073696.1 | - | 1 - 2 | 80 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271324.1 transcripts.
Set Membership: |
|

Mycbp
Genomic Map