Assay Details
| Gene Symbol: Tmed2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031722.2 | NP_113910.1 | 1 - 2 | 227 | 75 | |
| XM_006249327.2 | XP_006249389.1 | 1 - 2 | 276 | 75 | ||
| GenBank mRNA | BC062036.1 | - | 1 - 2 | 227 | 5623841 | 75 |
| X92097.1 | - | 1 - 2 | 198 | 75 | ||
More Information
Set Membership: |
|

Tmed2
Genomic Map