Assay Details
| Gene Symbol: Med30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130539.1 | NP_001124011.1 | 1 - 2 | 269 | 86 | |
| XM_006241646.2 | XP_006241708.1 | 1 - 2 | 277 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med30
Genomic Map