Assay Details
| Gene Symbol: Efnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107328.2 | NP_001100798.1 | 1 - 2 | 138 | 155 | |
| GenBank mRNA | DQ011669.1 | - | 1 - 2 | 138 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efnb2
Genomic Map