Assay Details
| Gene Symbol: Etv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106450.1 | NP_001099920.1 | 1 - 2 | 60 | 65 | |
| XM_006232620.2 | XP_006232682.1 | 2 - 3 | 151 | 65 | ||
| XM_006232621.2 | XP_006232683.1 | 2 - 3 | 768 | 65 | ||
| XM_008761141.1 | XP_008759363.1 | 2 - 3 | 470 | 65 | ||
| GenBank mRNA | BC168842.1 | - | 1 - 2 | 68 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etv3
Genomic Map