Assay Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106964.1 | NP_001100434.1 | 1514 | 62 | ||
| GenBank mRNA | BC167084.1 | - | 1609 | 62 | ||
| FQ232073.1 | - | 2 - 3 | 192 | 62 | ||
| FQ233310.1 | - | 2 - 3 | 192 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter
proteolysis
sensory perception of sound
biological_process
histone deacetylation
proteasome-mediated ubiquitin-dependent protein catabolic process
response to estrogen
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to steroid hormone
canonical Wnt signaling pathway
molecular_function
DNA binding
chromatin binding
transcription corepressor activity
beta-catenin binding
protein C-terminus binding
transcription factor binding
protein domain specific binding
histone binding
transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter
proteolysis
sensory perception of sound
biological_process
histone deacetylation
proteasome-mediated ubiquitin-dependent protein catabolic process
response to estrogen
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to steroid hormone
canonical Wnt signaling pathway
molecular_function
DNA binding
chromatin binding
transcription corepressor activity
beta-catenin binding
protein C-terminus binding
transcription factor binding
protein domain specific binding
histone binding
transcription regulatory region DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P079920 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P079917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179990 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079920 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079915 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079920 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079915 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P079915 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P079920 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1024714 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P900324 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75152 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46486 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10933765 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933766 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933764 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933756 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933759 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933761 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933757 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933763 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933762 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933758 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933760 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933767 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933768 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10933755 | - | - |
| Rat Genome 230 2.0 Array | 1379469_at | - | - |
| Rat Genome 230 2.0 Array | 1388478_at | - | - |
| Rat Genome 230 2.0 Array | 1388478_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393491_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379469_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393491_at | - | - |
| Affymetrix HT Rat Focus Array | 1393491_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379469_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379469_at | - | - |
| Affymetrix HT Rat Focus Array | 1393491_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00002894.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002895.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002898.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002899.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002897.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002902.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002900.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002896.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002890.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002904.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002903.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002893.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002906.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002901.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002907.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00002905.rn.1 | TC0X00000425.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388478_at | - | - |
| Rat Expression Set 230 | 1388478_PM_at | - | - |
| Rat Expression Set 230 | 1393491_PM_at | - | - |
| Rat Expression Set 230 | 1379469_PM_at | - | - |
| Rat Expression Set 230 | 1393491_at | - | - |
| Rat Expression Set 230 | 1379469_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933755 | - | - |
| Rat Genome U34 Set | rc_AA955760_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365469 | - | - |

Tbl1x
Genomic Map