Assay Details
| Gene Symbol: Sp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012761.1 | NP_036893.1 | 1 - 2 | 149 | 64 | |
| GenBank mRNA | U07610.1 | - | 1 - 2 | 149 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_012761.1, U07610.1 transcripts.
Set Membership: |
|

Sp4
Genomic Map