Assay Details
| Gene Symbol: Cnih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025132.1 | NP_001020303.1 | 3 - 4 | 444 | 87 | |
| XM_008760153.1 | XP_008758375.1 | 2 - 3 | 1022 | 87 | ||
| GenBank mRNA | BC091325.1 | - | 3 - 4 | 444 | 7313787 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|

Cnih2
Genomic Map