Assay Details
| Gene Symbol: Tmem182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109305.1 | NP_001102775.1 | 4 - 5 | 621 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P140873 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P670825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140854 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P670825 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140873 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P670825 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140873 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P670825 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P140873 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104649 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10922929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922927 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922930 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10922923 | - | - |
| Rat Genome 230 2.0 Array | 1385201_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385201_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900004491.rn.1 | TC0900000546.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385201_at | - | - |
| Rat Expression Set 230 | 1385201_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20881614 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351930 | - | - |

Tmem182
Genomic Map