Assay Details
| Gene Symbol: Tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC078715.1 | - | 1 - 2 | 210 | 7127457 | 67 |
| FQ224957.1 | - | 1 - 2 | 251 | 67 | ||
| FQ230650.1 | - | 1 - 2 | 250 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tomm20
Genomic Map