Assay Details
| Gene Symbol: Taf5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107442.1 | NP_001100912.1 | 1 - 2 | 183 | 146 | |
| XM_008772615.1 | XP_008770837.1 | 1 - 2 | 225 | 146 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107442.1, XM_008772615.1 transcripts.
Set Membership: |
|

Taf5l
Genomic Map