Assay Details
| Gene Symbol: Gpr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152242.2 | NP_689448.2 | 6 - 7 | 1193 | 61 | |
| XM_006255066.2 | XP_006255128.1 | 6 - 7 | 2245 | 61 | ||
| XM_008772264.1 | XP_008770486.1 | 6 - 7 | 1395 | 61 | ||
| XM_008772265.1 | XP_008770487.1 | 7 - 8 | 1659 | 61 | ||
| XM_008772266.1 | XP_008770488.1 | 6 - 7 | 2905 | 61 | ||
| XM_008772267.1 | XP_008770489.1 | 6 - 7 | 3168 | 61 | ||
| XM_008772268.1 | XP_008770490.1 | 7 - 8 | 1492 | 61 | ||
| XM_008772269.1 | XP_008770491.1 | 6 - 7 | 2274 | 61 | ||
| XM_008772270.1 | XP_008770492.1 | 6 - 7 | 2750 | 61 | ||
| GenBank mRNA | AF529886.1 | - | 6 - 7 | 1194 | 61 | |
| CV116515.1 | - | 4 - 5 | 649 | 61 | ||
| EX488169.1 | - | 3 - 4 | 304 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr56
Genomic Map