Assay Details
| Gene Symbol: Ube2v1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110345.2 | NP_001103815.2 | 1 - 2 | 63 | 85 | |
| GenBank mRNA | CO405669.1 | - | 1 - 2 | 38 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001110345.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
error-free postreplication DNA repair
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of NF-kappaB transcription factor activity
protein K63-linked ubiquitination
molecular_function
ubiquitin conjugating enzyme binding
ubiquitin conjugating enzyme activity
biological_process
error-free postreplication DNA repair
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of NF-kappaB transcription factor activity
protein K63-linked ubiquitination
molecular_function
ubiquitin conjugating enzyme binding
ubiquitin conjugating enzyme activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054719 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012262 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000410 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P045980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P058946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153962 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000410 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054719 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012262 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058946 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058946 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054719 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000410 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012262 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P045980 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054719 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012262 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000410 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P058946 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P102845 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59497 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10775311 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775310 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10775309 | - | - |
| Rat Genome 230 2.0 Array | 1398897_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395426_at | - | - |
| Rat Genome 230 2.0 Array | 1385820_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385820_at | - | - |
| Rat Genome 230 2.0 Array | 1398897_at | - | - |
| Rat Genome 230 2.0 Array | 1393991_PM_x_at | - | - |
| Rat Genome 230 2.0 Array | 1395426_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393991_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1395426_at | - | - |
| Affymetrix HT Rat Focus Array | 1398897_at | - | - |
| Affymetrix HT Rat Focus Array | 1398897_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395426_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300037080.rn.1 | TC0300004828.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037082.rn.1 | TC0300004828.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037081.rn.1 | TC0300004828.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398897_at | - | - |
| Rat Expression Set 230 | 1398897_PM_at | - | - |
| Rat Expression Set 230 | 1395426_PM_at | - | - |
| Rat Expression Set 230 | 1395426_at | - | - |
| Rat Expression Set 230 | 1385820_at | - | - |
| Rat Expression Set 230 | 1385820_PM_at | - | - |
| Rat Expression Set 230 | 1393991_PM_x_at | - | - |
| Rat Expression Set 230 | 1393991_x_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775309 | - | - |
| Rat Genome U34 Set | rc_AI169378_at | - | - |
| Rat Genome U34 Set | rc_AI229251_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357056 | - | - |

Ube2v1
Genomic Map