Assay Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271109.1 | NP_001258038.1 | 6 - 7 | 1053 | 112 | |
| NM_001271110.1 | NP_001258039.1 | 6 - 7 | 1081 | 112 | ||
| NM_080479.2 | NP_536727.2 | 6 - 7 | 1053 | 112 | ||
| XM_006237924.2 | XP_006237986.1 | 6 - 7 | 1143 | 112 | ||
| GenBank mRNA | AJ293617.1 | - | 5 - 6 | 994 | 112 | |
| AY724507.1 | - | 6 - 7 | 1066 | 112 | ||
| BC086998.1 | - | 5 - 6 | 1684 | 112 | ||
| BC107467.1 | - | 6 - 7 | 1053 | 7459969 | 112 | |
| FQ229095.1 | - | 6 - 7 | 1066 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Maged2
Genomic Map