Assay Details
| Gene Symbol: Wtip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759260.1 | XP_008757482.1 | 3 - 4 | 270 | 75 | |
| GenBank mRNA | BC167052.1 | - | 3 - 4 | 285 | 75 | |
More Information
Set Membership: |
|

Wtip
Genomic Map