Assay Details
| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276470.1 | NP_001263399.1 | 2 - 3 | 1093 | 74 | |
| XM_006230308.2 | XP_006230370.1 | 2 - 3 | 1172 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prrt2
Genomic Map