Assay Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108941.1 | NP_001102411.1 | 14 - 15 | 1640 | 73 | |
| XM_006232190.2 | XP_006232252.1 | 13 - 14 | 1533 | 73 | ||
| XM_008760889.1 | XP_008759111.1 | 14 - 15 | 2396 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to dietary excess
regulation of transcription from RNA polymerase II promoter
biological_process
regulation of gene expression
lipid catabolic process
histone deacetylation
regulation of cAMP metabolic process
multicellular organism growth
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
white fat cell differentiation
canonical Wnt signaling pathway
adipose tissue development
fat pad development
regulation of triglyceride metabolic process
molecular_function
DNA binding
transcription corepressor activity
beta-catenin binding
histone binding
transcription regulatory region DNA binding
protein N-terminus binding
negative regulation of transcription from RNA polymerase II promoter
response to dietary excess
regulation of transcription from RNA polymerase II promoter
biological_process
regulation of gene expression
lipid catabolic process
histone deacetylation
regulation of cAMP metabolic process
multicellular organism growth
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
white fat cell differentiation
canonical Wnt signaling pathway
adipose tissue development
fat pad development
regulation of triglyceride metabolic process
molecular_function
DNA binding
transcription corepressor activity
beta-catenin binding
histone binding
transcription regulatory region DNA binding
protein N-terminus binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P119215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61702 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P119215 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P119215 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P119215 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P965649 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P119215 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75336 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40744 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10814490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814496 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814489 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814491 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814492 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814494 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814487 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814488 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814493 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814495 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814497 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10814484 | - | - |
| Rat Genome 230 2.0 Array | 1386070_at | - | - |
| Rat Genome 230 2.0 Array | 1394107_at | - | - |
| Rat Genome 230 2.0 Array | 1394079_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379309_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379309_at | - | - |
| Rat Genome 230 2.0 Array | 1386070_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394107_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394079_at | - | - |
| Rat Genome 230 2.0 Array | 1389545_at | - | - |
| Rat Genome 230 2.0 Array | 1389545_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379309_at | - | - |
| Affymetrix HT Rat Focus Array | 1394079_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394079_at | - | - |
| Affymetrix HT Rat Focus Array | 1379309_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389545_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386070_at | - | - |
| Affymetrix HT Rat Focus Array | 1386070_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389545_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200005286.rn.1 | TC0200000865.rn.1 | RTA TC |
| Rat Expression Set 230 | 1394079_at | - | - |
| Rat Expression Set 230 | 1389545_PM_at | - | - |
| Rat Expression Set 230 | 1394079_PM_at | - | - |
| Rat Expression Set 230 | 1386070_PM_at | - | - |
| Rat Expression Set 230 | 1386070_at | - | - |
| Rat Expression Set 230 | 1389545_at | - | - |
| Rat Expression Set 230 | 1379309_PM_at | - | - |
| Rat Expression Set 230 | 1394107_PM_at | - | - |
| Rat Expression Set 230 | 1394107_at | - | - |
| Rat Expression Set 230 | 1379309_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814484 | - | - |
| Rat Genome U34 Set | rc_AA859701_at | - | - |
| Rat Genome U34 Set | rc_AA944610_at | - | - |
| Rat Genome U34 Set | rc_AA944938_at | - | - |
| Rat Genome U34 Set | rc_AI170260_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20749612 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355266 | - | - |

Tbl1xr1
Genomic Map