Assay Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003709.2 | NP_001003709.1 | 2 - 3 | 394 | 61 | |
| XM_006250251.2 | XP_006250313.1 | 3 - 4 | 349 | 61 | ||
| GenBank mRNA | AB186051.1 | - | 2 - 3 | 312 | 61 | |
| AB441169.1 | - | 2 - 3 | 312 | 61 | ||
| BC087648.1 | - | 2 - 3 | 394 | 7310651 | 61 | |
| FQ212748.1 | - | 2 - 3 | 412 | 61 | ||
| FQ213023.1 | - | 2 - 3 | 385 | 61 | ||
| FQ213507.1 | - | 2 - 3 | 395 | 61 | ||
| FQ216057.1 | - | 2 - 3 | 389 | 61 | ||
| FQ219875.1 | - | 2 - 3 | 264 | 61 | ||
| FQ226625.1 | - | 2 - 3 | 407 | 61 | ||
| FQ232457.1 | - | 2 - 3 | 371 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufc1
Genomic Map