Assay Details
| Gene Symbol: Lypd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007727.1 | NP_001007728.1 | 1 - 2 | 356 | 73 | |
| GenBank mRNA | BC082032.1 | - | 1 - 2 | 356 | 7130639 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Lypd1
Genomic Map