Assay Details
| Gene Symbol: Luc7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108291.1 | NP_001101761.1 | 5 - 6 | 593 | 112 | |
| XM_006247155.2 | XP_006247217.1 | 5 - 6 | 585 | 112 | ||
| XM_006247156.2 | XP_006247218.1 | 5 - 6 | 584 | 112 | ||
| XM_008768078.1 | XP_008766300.1 | 5 - 6 | 584 | 112 | ||
| GenBank mRNA | FQ212347.1 | - | 5 - 6 | 566 | 112 | |
| FQ231407.1 | - | 3 - 4 | 243 | 112 | ||
| FQ231682.1 | - | 5 - 6 | 566 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Luc7l3
Genomic Map